This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GM12130 SIRNA Assays ›
Gene Symbol:
Gm12130
Gene Name:
predicted gene 12130
Gene Aliases:
OTTMUSG00000005424
Chromosome Location:
Chr.11: 38491548 - 38520045 on Build GRCm38
UniGene ID:
Mm.61144
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n519875

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 357.00
Your Price
Online offer: 296.31
296.3
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gm12130
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_040295.1 3 414
GenBank mRNA AK145334.1 - 3 384
CJ052615.1 - 3 416
DV652764.1 - 3 358
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BY735543.1 - 3 384 Chr. 11 - 38493327 - 38520013 [-] on Build GRCm38
GenBankmRNA DV646593.1 - 3 370 Chr. 11 - 38493344 - 38520001 [-] on Build GRCm38
GenBankmRNA DV659262.1 - 4 497 Chr. 11 - 38493336 - 38520002 [-] on Build GRCm38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header