This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GM12130 SIRNA Assays ›
Gene Symbol:
Gm12130
Gene Name:
predicted gene 12130
Gene Aliases:
OTTMUSG00000005424
Chromosome Location:
Chr.11: 38491548 - 38520045 on Build GRCm38
UniGene ID:
Mm.61144
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n519874

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gm12130
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_040295.1 2 258
GenBank mRNA AK145334.1 - 2 228
CJ052615.1 - 2 260
DV652764.1 - 2 202
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BY735543.1 - 2 228 Chr. 11 - 38493327 - 38520013 [-] on Build GRCm38
GenBankmRNA BY030412.1 - 2 232 Chr. 11 - 38493544 - 38520013 [-] on Build GRCm38
GenBankmRNA DV646593.1 - 2 214 Chr. 11 - 38493344 - 38520001 [-] on Build GRCm38
GenBankmRNA DV659262.1 - 3 341 Chr. 11 - 38493336 - 38520002 [-] on Build GRCm38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0