This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other 4933431E20RIK SIRNA Assays ›
Gene Symbol:
4933431E20Rik
Gene Name:
RIKEN cDNA 4933431E20 gene
Gene Aliases:
-
Chromosome Location:
Chr.3: 107888850 - 107896213 on Build GRCm38
UniGene ID:
Mm.270500
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n519519

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 4933431E20Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015459.1 3 999
GenBank mRNA AK030251.1 - 4 622
AK036227.1 - 3 572
AK043835.1 - 3 1617
AK147824.1 - 4 596
BC060136.1 - 3 999
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BB617226.1 - 4 621 Chr. 3 - 107893199 - 107895766 [-] on Build GRCm38
GenBankmRNA BB629026.1 - 3 572 Chr. 3 - 107893165 - 107895798 [-] on Build GRCm38
GenBankmRNA CN536917.1 - 2 771 Chr. 3 - 107893223 - 107894574 [-] on Build GRCm38
GenBankmRNA BY127949.1 - 1 51 Chr. 3 - 107892973 - 107893308 [-] on Build GRCm38
GenBankmRNA CB722846.1 - 5 760 Chr. 3 - 107893223 - 107895774 [-] on Build GRCm38
GenBankmRNA AI509305.1 - 1 114 Chr. 3 - 107892959 - 107893368 [-] on Build GRCm38
GenBankmRNA BQ935019.1 - 2 511 Chr. 3 - 107892864 - 107894311 [-] on Build GRCm38
GenBankmRNA CX238834.1 - 4 696 Chr. 3 - 107893132 - 107895778 [-] on Build GRCm38
GenBankmRNA AA182357.1 - 1 114 Chr. 3 - 107892898 - 107893371 [-] on Build GRCm38
GenBankmRNA NM_177858.1 - 572 Chr. - - [-] on Build
GenBankmRNA BU504695.1 - 2 406 Chr. 3 - 107892753 - 107894206 [-] on Build GRCm38
GenBankmRNA BB269235.2 - 2 298 Chr. 3 - 107892897 - 107894098 [-] on Build GRCm38
GenBankmRNA BY740971.1 - 4 596 Chr. 3 - 107893114 - 107895741 [-] on Build GRCm38

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0