This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SNHG17 SIRNA Assays ›
Gene Symbol:
Snhg17
Gene Name:
small nucleolar RNA host gene 17
Gene Aliases:
9430008C03Rik
Chromosome Location:
Chr.2: 158353700 - 158361522 on Build GRCm38
UniGene ID:
Mm.119198
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n517798

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Snhg17
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015463.1 2 326
NR_027887.1 2 213
GenBank mRNA AK007971.1 - 2 214
AK040893.1 - 2 205
AK046044.1 - 2 314
AK079109.1 - 2 214
BU696406.1 - 2 337
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BB636878.1 - 2 202 Chr. 2 - 158360194 - 158361511 [-] on Build GRCm38
GenBankmRNA BY275519.1 - 2 186 Chr. 2 - 158360412 - 158361492 [-] on Build GRCm38
GenBankmRNA BY214464.1 - 2 235 Chr. 2 - 158360867 - 158361542 [-] on Build GRCm38
GenBankmRNA BU052986.1 - 2 166 Chr. 2 - 158359247 - 158361475 [-] on Build GRCm38
GenBankmRNA AV484598.1 - 2 152 Chr. 2 - 158359183 - 158361463 [-] on Build GRCm38
GenBankmRNA AW740594.1 - 2 135 Chr. 2 - 158359239 - 158361432 [-] on Build GRCm38
GenBankmRNA BB626634.1 - 2 214 Chr. 2 - 158360194 - 158361522 [-] on Build GRCm38
GenBankmRNA BU694596.1 - 2 327 Chr. 2 - 158360463 - 158361510 [-] on Build GRCm38
GenBankmRNA BY707970.1 - 2 214 Chr. 2 - 158355958 - 158361522 [-] on Build GRCm38
GenBankmRNA AV476660.1 - 2 192 Chr. 2 - 158359285 - 158361502 [-] on Build GRCm38
GenBankmRNA BF233030.1 - 142 Chr. - - [-] on Build
GenBankmRNA BQ712999.1 - 2 210 Chr. 2 - 158355948 - 158361519 [-] on Build GRCm38
GenBankmRNA AI931533.1 - 2 185 Chr. 2 - 158360393 - 158361486 [-] on Build GRCm38
GenBankmRNA CD554299.1 - 1 42 Chr. 2 - 158360170 - 158361017 [-] on Build GRCm38
GenBankmRNA AV492592.1 - 2 158 Chr. 2 - 158359223 - 158361467 [-] on Build GRCm38
GenBankmRNA AV467209.1 - 2 285 Chr. 2 - 158359226 - 158361470 [-] on Build GRCm38
GenBankmRNA BB857030.1 - 2 214 Chr. 2 - 158360548 - 158361522 [-] on Build GRCm38
GenBankmRNA AV574094.1 - 2 188 Chr. 2 - 158360262 - 158361488 [-] on Build GRCm38
GenBankmRNA AA762868.1 - 2 322 Chr. 2 - 158360552 - 158361492 [-] on Build GRCm38

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header