This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GM20752 SIRNA Assays ›
Gene Symbol:
Gm20752 Gm11033
Gene Name:
predicted gene, 20752
predicted gene 11033
Gene Aliases:
-
ENSMUSG00000079011
Chromosome Location:
Chr.3: 159076364 - 159308080 on Build GRCm38
Chromosome Location:
-
UniGene ID:
Mm.491054
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID n517089

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gm20752
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_040750.1 5 1981
GenBank mRNA AK158712.1 - 5 1983
Gene Symbol: Gm11033
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK144838.1 - 3 1455

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0