This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GM10658 SIRNA Assays ›
Gene Symbol:
Gm10658
Gene Name:
predicted gene 10658
Gene Aliases:
ENSMUSG00000074284
Chromosome Location:
Chr.9: 57056897 - 57071966 on Build GRCm38
UniGene ID:
Mm.361900
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n516908

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gm10658
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_045886.1 1 304
GenBank mRNA AK085949.1 - 1 295
AK143982.1 - 1 146
AK162906.1 - 1 305
BU696022.1 - 1 259
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BF123048.1 - 252 Chr. - - [-] on Build
GenBankmRNA BY732821.1 - 1 144 Chr. 9 - 57057327 - 57071806 [-] on Build GRCm38
GenBankmRNA BY076583.1 - 1 224 Chr. 9 - 57057694 - 57071885 [-] on Build GRCm38
GenBankmRNA BY724165.1 - 1 283 Chr. 9 - 57057486 - 57071945 [-] on Build GRCm38
GenBankmRNA BU695050.1 - 1 260 Chr. 9 - 57057635 - 57071921 [-] on Build GRCm38
GenBankmRNA AI789319.1 - 1 231 Chr. 9 - 57057580 - 57071893 [-] on Build GRCm38
GenBankmRNA BF471284.1 - 1 156 Chr. 9 - 57057568 - 57071818 [-] on Build GRCm38
GenBankmRNA BX516580.1 - 1 224 Chr. 9 - 57057580 - 57071886 [-] on Build GRCm38
GenBankmRNA BU697088.1 - 1 260 Chr. 9 - 57057607 - 57071921 [-] on Build GRCm38
GenBankmRNA BY086701.1 - 1 224 Chr. 9 - 57057703 - 57071885 [-] on Build GRCm38
GenBankmRNA CA491052.1 - 1 106 Chr. 9 - 57057225 - 57071733 [-] on Build GRCm38

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0