This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PMIS2 SIRNA Assays ›
Gene Symbol:
Pmis2
Gene Name:
Pmis2, sperm specific protein
Gene Aliases:
1700071L20Rik, 4930479M11Rik
Chromosome Location:
Chr.7: 30670722 - 30671605 on Build GRCm38
UniGene ID:
Mm.87326
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n516356

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 357.00
Your Price
Online offer: 296.31
296.3
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pmis2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027848.1 2 603
NR_027849.1 2 469
GenBank mRNA AK006932.1 - 2 471
AK015595.1 - 2 583
BC100424.1 - 2 589 30920315
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CA466270.1 - 2 458 Chr. 7 - 30670775 - 30671594 [-] on Build GRCm38
GenBankmRNA NM_183108.1 - 583 Chr. - - [-] on Build

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

single fertilization
binding of sperm to zona pellucida
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0