siRNA Details
Gene Symbol: LINC00664 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037194.1 | 11 | 1868 | |||
GenBank mRNA | BX537835.1 | - | 11 | 1868 |

Gene Symbol: LINC00664 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037194.1 | 11 | 1868 | |||
GenBank mRNA | BX537835.1 | - | 11 | 1868 |