This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LINC01029 SIRNA Assays ›
Gene Symbol:
LINC01029
Gene Name:
long intergenic non-protein coding RNA 1029
Gene Aliases:
-
Chromosome Location:
Chr.18: 77971294 - 77993727 on Build GRCh38
UniGene ID:
Hs.385670
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n514573

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 357.00
Your Price
Online offer: 296.31
296.3
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LINC01029
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_104127.1 2 777
GenBank mRNA BC016792.1 - 2 779

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0