siRNA Details
Gene Symbol: LINC00908 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015417.1 | 3 | 2188 | |||
GenBank mRNA | AK093936.1 | - | 3 | 2188 |

Gene Symbol: LINC00908 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015417.1 | 3 | 2188 | |||
GenBank mRNA | AK093936.1 | - | 3 | 2188 |