siRNA Details
Gene Symbol: LOC105371335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135171.1 | 3 | 303 | |||
NR_135172.1 | 2 | 181 | ||||
GenBank mRNA | HY013069.1 | - | 2 | 182 |

Gene Symbol: LOC105371335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135171.1 | 3 | 303 | |||
NR_135172.1 | 2 | 181 | ||||
GenBank mRNA | HY013069.1 | - | 2 | 182 |