siRNA Details
Gene Symbol: LOC100131315 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109767.1 | 1 | 92 | |||
GenBank mRNA | AK093166.1 | - | 1 | 234 | ||
AW582922.1 | - | 1 | 92 |

Gene Symbol: LOC100131315 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109767.1 | 1 | 92 | |||
GenBank mRNA | AK093166.1 | - | 1 | 234 | ||
AW582922.1 | - | 1 | 92 |