siRNA Details
Gene Symbol: LOC100506476 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109995.1 | 3 | 538 | |||
GenBank mRNA | CK822534.1 | - | 3 | 538 |

Gene Symbol: LOC100506476 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109995.1 | 3 | 538 | |||
GenBank mRNA | CK822534.1 | - | 3 | 538 |