siRNA Details
Gene Symbol: LOC102724814 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110555.1 | 1 | 165 | |||
GenBank mRNA | AK092855.1 | - | 1 | 165 |

Gene Symbol: LOC102724814 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110555.1 | 1 | 165 | |||
GenBank mRNA | AK092855.1 | - | 1 | 165 |