siRNA Details
Gene Symbol: RBPMS-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046205.1 | 3 | 614 | |||
GenBank mRNA | AK057935.1 | - | 3 | 616 | ||
BM845627.1 | - | 2 | 235 |

Gene Symbol: RBPMS-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046205.1 | 3 | 614 | |||
GenBank mRNA | AK057935.1 | - | 3 | 616 | ||
BM845627.1 | - | 2 | 235 |