This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RBPMS-AS1 SIRNA Assays ›
Gene Symbol:
RBPMS-AS1
Gene Name:
RBPMS antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.8: 30382119 - 30385401 on Build GRCh38
UniGene ID:
Hs.126813
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n511821

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RBPMS-AS1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_046205.1 2 306
GenBank mRNA AK057935.1 - 2 306
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DA680496.1 - 2 199 Chr. 8 - 30383130 - 30385293 [-] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0