This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OTUD6B-AS1 SIRNA Assays ›
Gene Symbol:
OTUD6B-AS1
Gene Name:
OTUD6B antisense RNA 1 (head to head)
Gene Aliases:
GS1-251I9.4
Chromosome Location:
Chr.8: 91059906 - 91070189 on Build GRCh38
UniGene ID:
Hs.153412
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n511705

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: OTUD6B-AS1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_110438.1 2 366
NR_110439.1 2 366
GenBank mRNA BC030623.1 - 2 371
BC105990.1 - 2 273
BM715864.1 - 2 300
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DA024402.1 - 2 267 Chr. 8 - 91069419 - 91070090 [-] on Build GRCh38
GenBankmRNA DA260430.1 - 2 257 Chr. 8 - 91069405 - 91070080 [-] on Build GRCh38
GenBankmRNA AB371438.1 - 2 230 Chr. 8 - 91069128 - 91070053 [-] on Build GRCh38
GenBankmRNA BF240725.1 - 221 Chr. - - [-] on Build
GenBankmRNA BG528387.1 - 2 299 Chr. 8 - 91063481 - 91070115 [-] on Build GRCh38
GenBankmRNA BF245340.1 - 218 Chr. - - [-] on Build
GenBankmRNA BF667646.1 - 216 Chr. - - [-] on Build
GenBankmRNA DA165632.1 - 2 185 Chr. 8 - 91069331 - 91070008 [-] on Build GRCh38
GenBankmRNA BM503514.1 - 2 149 Chr. 8 - 91069438 - 91069972 [-] on Build GRCh38
GenBankmRNA DA829556.1 - 2 262 Chr. 8 - 91069256 - 91070082 [-] on Build GRCh38
GenBankmRNA BI601380.1 - 2 221 Chr. 8 - 91069206 - 91070043 [-] on Build GRCh38
GenBankmRNA BF247188.1 - 2 218 Chr. 8 - 91069223 - 91070041 [-] on Build GRCh38
GenBankmRNA CB118521.1 - 2 254 Chr. 8 - 91069352 - 91070077 [-] on Build GRCh38
GenBankmRNA DB103066.1 - 2 215 Chr. 8 - 91069387 - 91070039 [-] on Build GRCh38
GenBankmRNA BG699467.1 - 264 Chr. - - [-] on Build
GenBankmRNA HY010814.1 - 2 216 Chr. 8 - 91069403 - 91070039 [-] on Build GRCh38
GenBankmRNA HY171161.1 - 2 231 Chr. 8 - 91069448 - 91070053 [-] on Build GRCh38
GenBankmRNA BU569421.1 - 2 294 Chr. 8 - 91069256 - 91070115 [-] on Build GRCh38
GenBankmRNA DA303246.1 - 2 268 Chr. 8 - 91069394 - 91070091 [-] on Build GRCh38
GenBankmRNA DA812941.1 - 2 216 Chr. 8 - 91063280 - 91070038 [-] on Build GRCh38
GenBankmRNA BF669699.1 - 2 221 Chr. 8 - 91069247 - 91070045 [-] on Build GRCh38
GenBankmRNA DB477823.1 - 2 216 Chr. 8 - 91069444 - 91070039 [-] on Build GRCh38
GenBankmRNA DA039495.1 - 2 207 Chr. 8 - 91069352 - 91070030 [-] on Build GRCh38
GenBankmRNA BQ632676.1 - 2 281 Chr. 8 - 91069351 - 91070102 [-] on Build GRCh38
GenBankmRNA BU172165.1 - 2 256 Chr. 8 - 91069195 - 91070079 [-] on Build GRCh38
GenBankmRNA AV725083.1 - 2 324 Chr. 8 - 91063522 - 91070021 [-] on Build GRCh38
GenBankmRNA DT220355.1 - 2 291 Chr. 8 - 91069397 - 91070113 [-] on Build GRCh38
GenBankmRNA BF030843.1 - 2 270 Chr. 8 - 91069301 - 91070095 [-] on Build GRCh38
GenBankmRNA DA447987.1 - 2 282 Chr. 8 - 91069436 - 91070105 [-] on Build GRCh38
GenBankmRNA CB120194.1 - 2 202 Chr. 8 - 91069387 - 91070025 [-] on Build GRCh38
GenBankmRNA HY165370.1 - 2 277 Chr. 8 - 91069473 - 91070100 [-] on Build GRCh38
GenBankmRNA BQ631803.1 - 2 211 Chr. 8 - 91069284 - 91070034 [-] on Build GRCh38
GenBankmRNA BM546159.1 - 2 216 Chr. 8 - 91068805 - 91070030 [-] on Build GRCh38
GenBankmRNA BM509004.1 - 2 271 Chr. 8 - 91069400 - 91070087 [-] on Build GRCh38
GenBankmRNA DB265743.1 - 2 207 Chr. 8 - 91069342 - 91070030 [-] on Build GRCh38
GenBankmRNA CB132486.1 - 2 197 Chr. 8 - 91069365 - 91070020 [-] on Build GRCh38
GenBankmRNA BY799587.2 - 2 230 Chr. 8 - 91069407 - 91070053 [-] on Build GRCh38
GenBankmRNA BP873123.1 - 2 241 Chr. 8 - 91069374 - 91070064 [-] on Build GRCh38
GenBankmRNA DB487816.1 - 2 216 Chr. 8 - 91069520 - 91070039 [-] on Build GRCh38
GenBankmRNA BF217211.1 - 2 219 Chr. 8 - 91069292 - 91070041 [-] on Build GRCh38
GenBankmRNA AL532758.3 - 2 217 Chr. 8 - 91068856 - 91070040 [-] on Build GRCh38
GenBankmRNA DB106765.1 - 2 225 Chr. 8 - 91069356 - 91070048 [-] on Build GRCh38
GenBankmRNA CD696748.1 - 2 296 Chr. 8 - 91069539 - 91070097 [-] on Build GRCh38
GenBankmRNA DA094126.1 - 2 200 Chr. 8 - 91063521 - 91070023 [-] on Build GRCh38
GenBankmRNA DB481667.1 - 2 199 Chr. 8 - 91069434 - 91070022 [-] on Build GRCh38
GenBankmRNA DB452795.1 - 2 216 Chr. 8 - 91069451 - 91070039 [-] on Build GRCh38
GenBankmRNA AA057559.1 - 2 196 Chr. 8 - 91069513 - 91070019 [-] on Build GRCh38
GenBankmRNA BF577021.1 - 215 Chr. - - [-] on Build
GenBankmRNA DA033391.1 - 2 216 Chr. 8 - 91069358 - 91070039 [-] on Build GRCh38
GenBankmRNA HY171260.1 - 2 290 Chr. 8 - 91069517 - 91070111 [-] on Build GRCh38
GenBankmRNA DB454695.1 - 2 183 Chr. 8 - 91069429 - 91070006 [-] on Build GRCh38
GenBankmRNA BU570220.1 - 2 233 Chr. 8 - 91069349 - 91070053 [-] on Build GRCh38
GenBankmRNA BU853047.1 - 2 210 Chr. 8 - 91069221 - 91070033 [-] on Build GRCh38
GenBankmRNA DB173611.1 - 2 165 Chr. 8 - 91069309 - 91069988 [-] on Build GRCh38
GenBankmRNA CB177477.1 - 2 282 Chr. 8 - 91069519 - 91070102 [-] on Build GRCh38
GenBankmRNA BE972763.1 - 2 242 Chr. 8 - 91069198 - 91070060 [-] on Build GRCh38
GenBankmRNA HY146757.1 - 2 231 Chr. 8 - 91069465 - 91070053 [-] on Build GRCh38
GenBankmRNA BX502356.1 - 2 217 Chr. 8 - 91069429 - 91070039 [-] on Build GRCh38
GenBankmRNA DA255024.1 - 2 289 Chr. 8 - 91069456 - 91070111 [-] on Build GRCh38
GenBankmRNA DN914588.1 - 2 203 Chr. 8 - 91069308 - 91070025 [-] on Build GRCh38
GenBankmRNA BU569666.1 - 2 220 Chr. 8 - 91063510 - 91070043 [-] on Build GRCh38
GenBankmRNA AV751156.1 - 2 187 Chr. 8 - 91069424 - 91070010 [-] on Build GRCh38
GenBankmRNA BM152347.1 - 2 231 Chr. 8 - 91069565 - 91070053 [-] on Build GRCh38
GenBankmRNA BU188109.1 - 2 233 Chr. 8 - 91069151 - 91070053 [-] on Build GRCh38
GenBankmRNA DA159897.1 - 2 291 Chr. 8 - 91069428 - 91070114 [-] on Build GRCh38
GenBankmRNA DN915989.1 - 2 312 Chr. 8 - 91069512 - 91070109 [-] on Build GRCh38
GenBankmRNA BU952250.1 - 2 206 Chr. 8 - 91069263 - 91070026 [-] on Build GRCh38
GenBankmRNA BI599069.1 - 2 221 Chr. 8 - 91069231 - 91070043 [-] on Build GRCh38
GenBankmRNA BU955253.1 - 233 Chr. - - [-] on Build
GenBankmRNA DA449032.1 - 2 230 Chr. 8 - 91069397 - 91070053 [-] on Build GRCh38
GenBankmRNA BJ997080.1 - 2 292 Chr. 8 - 91069568 - 91070115 [-] on Build GRCh38
GenBankmRNA CD107059.1 - 2 237 Chr. 8 - 91069053 - 91070040 [-] on Build GRCh38
GenBankmRNA DB447119.1 - 2 216 Chr. 8 - 91069513 - 91070039 [-] on Build GRCh38
GenBankmRNA BU160824.1 - 2 233 Chr. 8 - 91069175 - 91070053 [-] on Build GRCh38
GenBankmRNA BM352418.1 - 2 289 Chr. 8 - 91069420 - 91070102 [-] on Build GRCh38
GenBankmRNA DA156972.1 - 2 196 Chr. 8 - 91069334 - 91070019 [-] on Build GRCh38
GenBankmRNA DA130457.1 - 2 177 Chr. 8 - 91069334 - 91069999 [-] on Build GRCh38
GenBankmRNA BU159364.1 - 2 197 Chr. 8 - 91069016 - 91070020 [-] on Build GRCh38
GenBankmRNA BM905024.1 - 241 Chr. - - [-] on Build
GenBankmRNA DB472212.1 - 2 216 Chr. 8 - 91069433 - 91070039 [-] on Build GRCh38
GenBankmRNA DB482984.1 - 2 227 Chr. 8 - 91069443 - 91070050 [-] on Build GRCh38
GenBankmRNA T35512.1 - 2 195 Chr. 8 - 91069528 - 91070018 [-] on Build GRCh38
GenBankmRNA DR762734.1 - 2 206 Chr. 8 - 91069078 - 91070029 [-] on Build GRCh38
GenBankmRNA BG290609.1 - 2 196 Chr. 8 - 91069094 - 91070019 [-] on Build GRCh38
GenBankmRNA BF574970.1 - 2 216 Chr. 8 - 91069240 - 91070039 [-] on Build GRCh38
GenBankmRNA DA840239.1 - 2 216 Chr. 8 - 91069385 - 91070039 [-] on Build GRCh38
GenBankmRNA HY174754.1 - 2 217 Chr. 8 - 91069434 - 91070039 [-] on Build GRCh38
GenBankmRNA BI759621.1 - 2 165 Chr. 8 - 91069173 - 91069988 [-] on Build GRCh38
GenBankmRNA BQ230179.1 - 2 268 Chr. 8 - 91069165 - 91070091 [-] on Build GRCh38
GenBankmRNA BX957028.1 - 2 218 Chr. 8 - 91069303 - 91070035 [-] on Build GRCh38
GenBankmRNA DA121039.1 - 2 232 Chr. 8 - 91063571 - 91070054 [-] on Build GRCh38
GenBankmRNA BI545472.1 - 2 221 Chr. 8 - 91069257 - 91070043 [-] on Build GRCh38
GenBankmRNA BF541012.1 - 2 274 Chr. 8 - 91069327 - 91070097 [-] on Build GRCh38
GenBankmRNA BG778864.1 - 2 231 Chr. 8 - 91069247 - 91070053 [-] on Build GRCh38
GenBankmRNA DA317375.1 - 2 289 Chr. 8 - 91063596 - 91070112 [-] on Build GRCh38
GenBankmRNA CN387718.1 - 2 215 Chr. 8 - 91069177 - 91070038 [-] on Build GRCh38
GenBankmRNA DA272628.1 - 2 227 Chr. 8 - 91069375 - 91070050 [-] on Build GRCh38
GenBankmRNA CB052237.1 - 2 216 Chr. 8 - 91063691 - 91070032 [-] on Build GRCh38
GenBankmRNA BG502674.1 - 2 234 Chr. 8 - 91069213 - 91070056 [-] on Build GRCh38
GenBankmRNA BI912732.1 - 2 209 Chr. 8 - 91069140 - 91070033 [-] on Build GRCh38
GenBankmRNA AV718180.1 - 2 324 Chr. 8 - 91069333 - 91070147 [-] on Build GRCh38
GenBankmRNA HY047845.1 - 2 216 Chr. 8 - 91063649 - 91070039 [-] on Build GRCh38
GenBankmRNA BF207734.1 - 2 217 Chr. 8 - 91069214 - 91070039 [-] on Build GRCh38
GenBankmRNA DB469881.1 - 2 216 Chr. 8 - 91069490 - 91070039 [-] on Build GRCh38
GenBankmRNA DB465278.1 - 2 183 Chr. 8 - 91069461 - 91070006 [-] on Build GRCh38
GenBankmRNA BE738150.1 - 194 Chr. - - [-] on Build
GenBankmRNA CB107298.1 - 2 177 Chr. 8 - 91063695 - 91070000 [-] on Build GRCh38
GenBankmRNA BM792763.1 - 1 17 Chr. 8 - 91063519 - 91069732 [-] on Build GRCh38
GenBankmRNA AL601160.1 - 2 214 Chr. 8 - 91069502 - 91070036 [-] on Build GRCh38
GenBankmRNA DB482656.1 - 2 258 Chr. 8 - 91063641 - 91070080 [-] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0