This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LOC101929450 SIRNA Assays ›
Gene Symbol:
LOC101929450
Gene Name:
uncharacterized LOC101929450
Gene Aliases:
-
Chromosome Location:
Chr.8: 29748309 - 29798492 on Build GRCh38
UniGene ID:
Hs.512440
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n511600

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC101929450
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_125814.1 1 82
NR_125815.1 1 82
NR_125816.1 1 82
GenBank mRNA BC015784.1 - 1 37
BC047326.1 - 1 41
BC082237.1 - 1 66
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BG763798.1 - 1 67 Chr. 8 - 29748325 - 29797001 [+] on Build GRCh38
GenBankmRNA BG763740.1 - 1 38 Chr. 8 - 29748354 - 29791578 [+] on Build GRCh38
GenBankmRNA BG766806.1 - 1 67 Chr. 8 - 29748325 - 29796934 [+] on Build GRCh38
GenBankmRNA BG764943.1 - 1 39 Chr. 8 - 29748354 - 29791663 [+] on Build GRCh38
GenBankmRNA BM472308.1 - 1 83 Chr. 8 - 29748309 - 29750163 [+] on Build GRCh38
GenBankmRNA BM453889.1 - 1 41 Chr. 8 - 29748350 - 29791585 [+] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0