siRNA Details
Gene Symbol: LOC100506107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134683.1 | 2 | 265 | |||
GenBank mRNA | AF086144.1 | - | 2 | 265 | ||
BU078148.1 | - | 2 | 258 |

Gene Symbol: LOC100506107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134683.1 | 2 | 265 | |||
GenBank mRNA | AF086144.1 | - | 2 | 265 | ||
BU078148.1 | - | 2 | 258 |