This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CARMN SIRNA Assays ›
Gene Symbol:
CARMN
Gene Name:
cardiac mesoderm enhancer-associated non-coding RNA
Gene Aliases:
CARMEN, MIR143HG
Chromosome Location:
Chr.5: 149406845 - 149432836 on Build GRCh38
UniGene ID:
Hs.519666
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n510766

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CARMN
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_105059.1 1 134
NR_105060.1 1 134
GenBank mRNA AK126481.1 - 1 101
BX640700.1 - 1 17
CR933654.1 - 1 100
HQ848934.1 - 1 134
HQ848935.1 - 1 134
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DA325631.1 - 1 16 Chr. 5 - 149406963 - 149424464 [+] on Build GRCh38
GenBankmRNA HY004392.1 - 1 102 Chr. 5 - 149406878 - 149424291 [+] on Build GRCh38
GenBankmRNA DA892159.1 - 1 99 Chr. 5 - 149406880 - 149424330 [+] on Build GRCh38
GenBankmRNA DB280782.1 - 1 16 Chr. 5 - 149406963 - 149424396 [+] on Build GRCh38
GenBankmRNA HY023115.1 - 1 102 Chr. 5 - 149406878 - 149423277 [+] on Build GRCh38
GenBankmRNA DB232103.1 - 1 16 Chr. 5 - 149406963 - 149421941 [+] on Build GRCh38
GenBankmRNA DA338115.1 - 1 103 Chr. 5 - 149406877 - 149424346 [+] on Build GRCh38
GenBankmRNA BM716826.1 - 1 62 Chr. 5 - 149406916 - 149425229 [+] on Build GRCh38
GenBankmRNA BM701454.1 - 1 63 Chr. 5 - 149406916 - 149424254 [+] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0