siRNA Details
Gene Symbol: LOC101929544 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134273.1 | 2 | 660 | |||
NR_134274.1 | 2 | 389 | ||||
NR_134275.1 | 2 | 253 |

Gene Symbol: LOC101929544 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134273.1 | 2 | 660 | |||
NR_134274.1 | 2 | 389 | ||||
NR_134275.1 | 2 | 253 |