siRNA Details
Gene Symbol: LOC286059 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038873.1 | 3 | 947 | |||
GenBank mRNA | AK024295.1 | - | 3 | 968 |

Gene Symbol: LOC286059 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038873.1 | 3 | 947 | |||
GenBank mRNA | AK024295.1 | - | 3 | 968 |