siRNA Details
Gene Symbol: LINC00971 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033860.1 | 11 | 1726 | |||
GenBank mRNA | BC068246.1 | - | 11 | 1728 |

Gene Symbol: LINC00971 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033860.1 | 11 | 1726 | |||
GenBank mRNA | BC068246.1 | - | 11 | 1728 |