siRNA Details
Gene Symbol: LINC00969 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128346.1 | - | 2 | 2621 | ||
BC144457.1 | - | 2 | 2152 | |||
BG983179.1 | - | 1, 2 | 200 |

Gene Symbol: LINC00969 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128346.1 | - | 2 | 2621 | ||
BC144457.1 | - | 2 | 2152 | |||
BG983179.1 | - | 1, 2 | 200 |