siRNA Details
Gene Symbol: SUCLG2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109992.1 | 4 | 1465 | |||
GenBank mRNA | AL832184.1 | - | 4 | 1405 |

Gene Symbol: SUCLG2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109992.1 | 4 | 1465 | |||
GenBank mRNA | AL832184.1 | - | 4 | 1405 |