siRNA Details
Gene Symbol: LINC00877 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104116.1 | 5 | 1043 | |||
GenBank mRNA | AK097190.1 | - | 5 | 1015 |

Gene Symbol: LINC00877 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104116.1 | 5 | 1043 | |||
GenBank mRNA | AK097190.1 | - | 5 | 1015 |