siRNA Details
Gene Symbol: LINC01625 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033919.1 | 4 | 1092 | |||
GenBank mRNA | AK123801.1 | - | 4 | 1091 |

Gene Symbol: LINC01625 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033919.1 | 4 | 1092 | |||
GenBank mRNA | AK123801.1 | - | 4 | 1091 |