siRNA Details
Gene Symbol: LINC01352 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110797.1 | 4 | 691 | |||
GenBank mRNA | BC034007.1 | - | 4 | 696 | ||
BC045735.1 | - | 4 | 691 |

Gene Symbol: LINC01352 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110797.1 | 4 | 691 | |||
GenBank mRNA | BC034007.1 | - | 4 | 696 | ||
BC045735.1 | - | 4 | 691 |