This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF503-AS2 SIRNA Assays ›
Gene Symbol:
ZNF503-AS2
Gene Name:
ZNF503 antisense RNA 2
Gene Aliases:
C10orf41, NCRNA00245
Chromosome Location:
Chr.10: 75401528 - 75408982 on Build GRCh38
UniGene ID:
Hs.534598
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n508796

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF503-AS2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024421.1 2 772
NR_024422.1 2 340
NR_110299.1 2 336
NR_110300.1 2 108
NR_110301.1 2 225
NR_110302.1 2 158
GenBank mRNA BC017426.1 - 2 340
BC029963.1 - 2 108
BE392199.1 - 2 227
BJ990170.1 - 2 340
DA634630.1 - 2 160
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA N79147.1 - 2 185 Chr. 10 - 75403917 - 75407344 [+] on Build GRCh38
GenBankmRNA DA135297.1 - 2 255 Chr. 10 - 75402045 - 75407589 [+] on Build GRCh38
GenBankmRNA N78550.1 - 2 183 Chr. 10 - 75403917 - 75407357 [+] on Build GRCh38
GenBankmRNA BX344557.2 - 2 401 Chr. 10 - 75401898 - 75407941 [+] on Build GRCh38
GenBankmRNA CN484197.1 - 2 392 Chr. 10 - 75401919 - 75407527 [+] on Build GRCh38
GenBankmRNA HY127970.1 - 2 381 Chr. 10 - 75401920 - 75407397 [+] on Build GRCh38
GenBankmRNA BX400778.2 - 2 401 Chr. 10 - 75401898 - 75407779 [+] on Build GRCh38
GenBankmRNA BF343288.1 - 165 Chr. - - [-] on Build
GenBankmRNA AA426393.1 - 2 85 Chr. 10 - 75404011 - 75407449 [+] on Build GRCh38
GenBankmRNA BG542799.1 - 2 337 Chr. 10 - 75403054 - 75407673 [+] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header