This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HAND2-AS1 SIRNA Assays ›
Gene Symbol:
HAND2-AS1
Gene Name:
HAND2 antisense RNA 1 (head to head)
Gene Aliases:
DEIN, NBLA00301
Chromosome Location:
Chr.4: 173528600 - 173591255 on Build GRCh38
UniGene ID:
Hs.61435
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n508794

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: HAND2-AS1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003679.2 1 61
NR_136192.1 1 148
NR_136193.1 1 148
NR_136194.1 1 148
NR_136195.1 1 148
NR_136196.1 1 148
NR_136198.1 1 148
NR_136199.1 1 148
NR_136200.1 1 148
NR_136201.1 1 148
GenBank mRNA AB075501.1 - 1 60
AK054605.1 - 1 57
AU253586.1 - 1 57
BG519698.1 - 1 27
BQ882603.1 - 1 148
DA665500.1 - 1 57
DB292439.1 - 1 81
DC386781.1 - 1 59
EF529495.1 - 1 61
EF529496.1 - 1 61
EF529497.1 - 1 61
EF529498.1 - 1 61
EF529499.1 - 1 61
EF529500.1 - 1 61
EF529501.1 - 1 61
EF529502.1 - 1 61
EF529503.1 - 1 61
EF529504.1 - 1 61
EF529505.1 - 1 61
EF529506.1 - 1 61
EF529507.1 - 1 61
EF529508.1 - 1 61
EF529509.1 - 1 61
EU000853.1 - 1 61
EU000854.1 - 1 61
EU000855.1 - 1 61
EU000856.1 - 1 61
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DC360108.1 - 1 48 Chr. 4 - 173530472 - 173538277 [+] on Build GRCh38
GenBankmRNA DC386933.1 - 1 44 Chr. 4 - 173530475 - 173538159 [+] on Build GRCh38
GenBankmRNA DA614232.1 - 1 57 Chr. 4 - 173530462 - 173537219 [+] on Build GRCh38
GenBankmRNA BF304375.1 - 37 Chr. - - [-] on Build
GenBankmRNA DC354299.1 - 1 57 Chr. 4 - 173530462 - 173538157 [+] on Build GRCh38
GenBankmRNA DC360375.1 - 1 59 Chr. 4 - 173530460 - 173538107 [+] on Build GRCh38
GenBankmRNA DC360232.1 - 1 29 Chr. 4 - 173530491 - 173538106 [+] on Build GRCh38
GenBankmRNA DA927156.1 - 1 57 Chr. 4 - 173530462 - 173537450 [+] on Build GRCh38
GenBankmRNA BE312774.1 - 1 17 Chr. 4 - 173530502 - 173537295 [+] on Build GRCh38
GenBankmRNA BP379670.1 - 1 43 Chr. 4 - 173530475 - 173538344 [+] on Build GRCh38
GenBankmRNA DA919798.1 - 1 57 Chr. 4 - 173530462 - 173537054 [+] on Build GRCh38
GenBankmRNA DC386963.1 - 1 24 Chr. 4 - 173530495 - 173538165 [+] on Build GRCh38
GenBankmRNA DA606691.1 - 1 23 Chr. 4 - 173530496 - 173537315 [+] on Build GRCh38
GenBankmRNA DC360466.1 - 1 57 Chr. 4 - 173530462 - 173538130 [+] on Build GRCh38
GenBankmRNA DC297746.1 - 1 50 Chr. 4 - 173530469 - 173538168 [+] on Build GRCh38
GenBankmRNA DC381495.1 - 1 59 Chr. 4 - 173530460 - 173538307 [+] on Build GRCh38
GenBankmRNA DC381155.1 - 1 44 Chr. 4 - 173530475 - 173538158 [+] on Build GRCh38
GenBankmRNA DC360477.1 - 1 44 Chr. 4 - 173530475 - 173538162 [+] on Build GRCh38
GenBankmRNA BQ777900.1 - 1 129 Chr. 4 - 173530390 - 173538233 [+] on Build GRCh38
GenBankmRNA DC388584.1 - 1 57 Chr. 4 - 173530462 - 173538318 [+] on Build GRCh38
GenBankmRNA R24426.1 - 25 Chr. - - [-] on Build
GenBankmRNA DA823966.1 - 1 44 Chr. 4 - 173530475 - 173537465 [+] on Build GRCh38
GenBankmRNA DC361029.1 - 1 45 Chr. 4 - 173530475 - 173538187 [+] on Build GRCh38
GenBankmRNA AL523783.3 - 1 40 Chr. 4 - 173530479 - 173538777 [+] on Build GRCh38
GenBankmRNA CK902262.1 - 1 110 Chr. 4 - 173530409 - 173538202 [+] on Build GRCh38
GenBankmRNA AU252421.1 - 1 86 Chr. 4 - 173530462 - 173537325 [+] on Build GRCh38
GenBankmRNA AL522836.3 - 1 45 Chr. 4 - 173530474 - 173537415 [+] on Build GRCh38
GenBankmRNA BP380043.1 - 1 59 Chr. 4 - 173530460 - 173538314 [+] on Build GRCh38
GenBankmRNA DB249244.1 - 1 57 Chr. 4 - 173530462 - 173537046 [+] on Build GRCh38
GenBankmRNA DC360095.1 - 1 57 Chr. 4 - 173530462 - 173538260 [+] on Build GRCh38
GenBankmRNA DC360996.1 - 1 57 Chr. 4 - 173530462 - 173538121 [+] on Build GRCh38
GenBankmRNA DC360499.1 - 1 57 Chr. 4 - 173530462 - 173538149 [+] on Build GRCh38
GenBankmRNA DC354261.1 - 1 57 Chr. 4 - 173530462 - 173538142 [+] on Build GRCh38
GenBankmRNA DC360579.1 - 1 24 Chr. 4 - 173530495 - 173538334 [+] on Build GRCh38
GenBankmRNA DA922612.1 - 1 59 Chr. 4 - 173530461 - 173537218 [+] on Build GRCh38
GenBankmRNA DC380727.1 - 1 57 Chr. 4 - 173530462 - 173538322 [+] on Build GRCh38
GenBankmRNA BU146691.1 - 1 40 Chr. 4 - 173530479 - 173538629 [+] on Build GRCh38
GenBankmRNA DC361115.1 - 1 61 Chr. 4 - 173530458 - 173538164 [+] on Build GRCh38
GenBankmRNA DC360170.1 - 1 57 Chr. 4 - 173530462 - 173538097 [+] on Build GRCh38
GenBankmRNA DC298067.1 - 1 59 Chr. 4 - 173530460 - 173538137 [+] on Build GRCh38
GenBankmRNA DC361312.1 - 1 57 Chr. 4 - 173530462 - 173538169 [+] on Build GRCh38
GenBankmRNA DC380045.1 - 1 57 Chr. 4 - 173530462 - 173538250 [+] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0