siRNA Details
Gene Symbol: LINC01447 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108090.1 | 4 | 2724 | |||
GenBank mRNA | AK056440.1 | - | 4 | 2724 |

Gene Symbol: LINC01447 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108090.1 | 4 | 2724 | |||
GenBank mRNA | AK056440.1 | - | 4 | 2724 |