siRNA Details
Gene Symbol: LOC100507506 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103764.1 | 4 | 731 | |||
GenBank mRNA | AK092451.1 | - | 4 | 731 |

Gene Symbol: LOC100507506 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103764.1 | 4 | 731 | |||
GenBank mRNA | AK092451.1 | - | 4 | 731 |