siRNA Details
Gene Symbol: LINC01049 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120414.1 | 6 | 662 | |||
GenBank mRNA | BC038529.1 | - | 6 | 662 |

Gene Symbol: LINC01049 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120414.1 | 6 | 662 | |||
GenBank mRNA | BC038529.1 | - | 6 | 662 |