siRNA Details
Gene Symbol: LINC01450 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110831.1 | 3 | 2165 | |||
GenBank mRNA | BX647937.1 | - | 3 | 2166 |

Gene Symbol: LINC01450 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110831.1 | 3 | 2165 | |||
GenBank mRNA | BX647937.1 | - | 3 | 2166 |