siRNA Details
Gene Symbol: LOC340512 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126029.1 | 2 | 275 | |||
GenBank mRNA | AK097706.1 | - | 2 | 275 |

Gene Symbol: LOC340512 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126029.1 | 2 | 275 | |||
GenBank mRNA | AK097706.1 | - | 2 | 275 |