siRNA Details
Gene Symbol: LOC284798 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027091.1 | 4 | 2952 | |||
NR_027092.1 | 4 | 2245 | ||||
NR_027093.1 | 4 | 2352 | ||||
GenBank mRNA | AK095800.1 | - | 4 | 2952 |

Gene Symbol: LOC284798 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027091.1 | 4 | 2952 | |||
NR_027092.1 | 4 | 2245 | ||||
NR_027093.1 | 4 | 2352 | ||||
GenBank mRNA | AK095800.1 | - | 4 | 2952 |