siRNA Details
Gene Symbol: LOC101929592 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125986.1 | 3 | 248 | |||
GenBank mRNA | BX280555.1 | - | 3 | 248 |

Gene Symbol: LOC101929592 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125986.1 | 3 | 248 | |||
GenBank mRNA | BX280555.1 | - | 3 | 248 |