siRNA Details
Gene Symbol: LOC101927692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120628.1 | 3 | 916 | |||
GenBank mRNA | AK098198.1 | - | 3 | 916 |

Gene Symbol: LOC101927692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120628.1 | 3 | 916 | |||
GenBank mRNA | AK098198.1 | - | 3 | 916 |