siRNA Details
Gene Symbol: LOC101927394 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110131.1 | 4 | 1215 | |||
GenBank mRNA | AK124857.1 | - | 4 | 1215 |

Gene Symbol: LOC101927394 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110131.1 | 4 | 1215 | |||
GenBank mRNA | AK124857.1 | - | 4 | 1215 |