This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LRFN2 SIRNA Assays ›
Gene Symbol:
LRFN2
Gene Name:
leucine rich repeat and fibronectin type III domain containing 2
Gene Aliases:
FIGLER2, KIAA1246, SALM1
Chromosome Location:
Chr.6: 40391589 - 40587464 on Build GRCh38
UniGene ID:
Hs.250015
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n505153

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LRFN2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011514761.2 XP_011513063.1 2 193
XM_017011110.1 XP_016866599.1 2 1234
XM_017011111.1 XP_016866600.1 2 191

Back To Top

More Information


Gene Ontology Categories:

Function(s)

chemical synaptic transmission
axonogenesis

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0