This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LOC101928118 SIRNA Assays ›
Gene Symbol:
LOC101928118
Gene Name:
uncharacterized LOC101928118
Gene Aliases:
-
Chromosome Location:
Chr.1: 95163219 - 95233982 on Build GRCh38
UniGene ID:
Hs.656443
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n504990

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC101928118
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_125948.1 2, 3 434
NR_125949.1 2, 3 434
GenBank mRNA AK090700.1 - 2, 3 434
DB219127.1 - 2, 3 410
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DA058655.1 - 2, 3 434 Chr. 1 - 95165089 - 95233982 [-] on Build GRCh38
GenBankmRNA CD696937.1 - 2, 3 461 Chr. 1 - 95174124 - 95233981 [-] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0