siRNA Details
Gene Symbol: LINC01398 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121680.1 | 3 | 678 | |||
GenBank mRNA | BC021746.1 | - | 3 | 682 |

Gene Symbol: LINC01398 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121680.1 | 3 | 678 | |||
GenBank mRNA | BC021746.1 | - | 3 | 682 |