This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LOC643072 SIRNA Assays ›
Gene Symbol:
LOC643072
Gene Name:
uncharacterized LOC643072
Gene Aliases:
-
Chromosome Location:
Chr.2: 159615294 - 159618042 on Build GRCh38
UniGene ID:
Hs.632541
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n504285

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC643072
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_110587.1 2 330
NR_110588.1 2 327
GenBank mRNA DA209039.1 - 2 319
HY006825.1 - 2 331
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DA552768.1 - 2 311 Chr. 2 - 159615314 - 159616158 [+] on Build GRCh38
GenBankmRNA DA024324.1 - 2 302 Chr. 2 - 159615322 - 159616142 [+] on Build GRCh38
GenBankmRNA AL583527.3 - 2 270 Chr. 2 - 159615352 - 159616501 [+] on Build GRCh38
GenBankmRNA BI821448.1 - 2 303 Chr. 2 - 159615320 - 159616326 [+] on Build GRCh38
GenBankmRNA DA897587.1 - 2 327 Chr. 2 - 159615298 - 159616136 [+] on Build GRCh38
GenBankmRNA BM692242.1 - 2 224 Chr. 2 - 159615400 - 159616288 [+] on Build GRCh38
GenBankmRNA BM906171.1 - 386 Chr. - - [-] on Build

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-6f7678f454-w8g9k:80/100.66.128.142:80.
git-commit: 7b0558793d0e70b0085ba95ba375cdfe261f8cbf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0