This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LOC101929282 SIRNA Assays ›
Gene Symbol:
LOC101929282
Gene Name:
uncharacterized LOC101929282
Gene Aliases:
-
Chromosome Location:
Chr.2: 150628897 - 150635357 on Build GRCh38
UniGene ID:
Hs.126866
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n504244

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC101929282
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_110244.1 3 389
NR_110245.1 2 273
NR_110246.1 2 314
NR_110247.1 2 314
GenBank mRNA BF573640.1 - 2 215
BX100556.1 - 2 144
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AA421345.1 - 2 144 Chr. 2 - 150629067 - 150634868 [+] on Build GRCh38
GenBankmRNA DB060520.1 - 3 466 Chr. 2 - 150628897 - 150634716 [+] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header