This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MIR99AHG SIRNA Assays ›
Gene Symbol:
MIR99AHG
Gene Name:
mir-99a-let-7c cluster host gene
Gene Aliases:
C21orf34, C21orf35, LINC00478, MONC
Chromosome Location:
Chr.21: 16070488 - 16631727 on Build GRCh38
UniGene ID:
Hs.473394
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n503887

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 357.00
Your Price
Online offer: 296.31
296.3
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: MIR99AHG
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027790.3 5, 6 511
NR_027791.3 4, 5 429
NR_111005.2 5, 6 556
NR_136544.1 2, 3 581
NR_136545.1 2, 3 581
NR_136550.1 2, 3 320
NR_136551.1 4, 5 464
NR_136552.1 4, 5 464
GenBank mRNA AK095614.1 - 5, 6 511
AL700268.1 - 4, 5 340
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DA498845.1 - 5 511 Chr. 21 - 16070522 - 16487553 [+] on Build GRCh38
GenBankmRNA DA498833.1 - 5 511 Chr. 21 - 16070522 - 16487553 [+] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0