This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LINC00998 SIRNA Assays ›
Gene Symbol:
LINC00998
Gene Name:
long intergenic non-protein coding RNA 998
Gene Aliases:
-
Chromosome Location:
Chr.7: 113116718 - 113118582 on Build GRCh38
UniGene ID:
Hs.117929
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n503794

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 357.00
Your Price
Online offer: 296.31
296.3
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LINC00998
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024412.1 1 101
GenBank mRNA DN995392.1 - 1 75
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CN430142.1 - 1 60 Chr. 7 - 113117303 - 113118530 [-] on Build GRCh38
GenBankmRNA CR625886.1 - 1 118 Chr. 7 - 113116990 - 113118599 [-] on Build GRCh38
GenBankmRNA BE866300.1 - 1 57 Chr. 7 - 113117180 - 113118538 [-] on Build GRCh38
GenBankmRNA BG527392.1 - 1 74 Chr. 7 - 113117103 - 113118554 [-] on Build GRCh38
GenBankmRNA BG776999.1 - 1 75 Chr. 7 - 113117120 - 113118555 [-] on Build GRCh38
GenBankmRNA BG687464.1 - 1 74 Chr. 7 - 113117091 - 113118554 [-] on Build GRCh38
GenBankmRNA BG403932.1 - 1 38 Chr. 7 - 113117103 - 113118519 [-] on Build GRCh38
GenBankmRNA BG532857.1 - 1 74 Chr. 7 - 113117111 - 113118554 [-] on Build GRCh38
GenBankmRNA BI766819.1 - 1 56 Chr. 7 - 113116870 - 113118537 [-] on Build GRCh38
GenBankmRNA BG493956.1 - 1 77 Chr. 7 - 113117194 - 113118557 [-] on Build GRCh38
GenBankmRNA BX395677.2 - 1 118 Chr. 7 - 113117644 - 113118599 [-] on Build GRCh38
GenBankmRNA BM857697.1 - 1 45 Chr. 7 - 113117417 - 113118519 [-] on Build GRCh38
GenBankmRNA BG530236.1 - 1 74 Chr. 7 - 113117016 - 113118554 [-] on Build GRCh38
GenBankmRNA BG390172.1 - 53 Chr. - - [-] on Build
GenBankmRNA BG390022.1 - 1 53 Chr. 7 - 113117257 - 113118534 [-] on Build GRCh38
GenBankmRNA BP421775.1 - 1 36 Chr. 7 - 113117268 - 113118517 [-] on Build GRCh38
GenBankmRNA BP421774.1 - 1 37 Chr. 7 - 113117279 - 113118517 [-] on Build GRCh38
GenBankmRNA AW967865.1 - 1 47 Chr. 7 - 113117261 - 113118528 [-] on Build GRCh38
GenBankmRNA BG532952.1 - 1 74 Chr. 7 - 113117052 - 113118554 [-] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0