siRNA Details
Gene Symbol: LINC01602 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130934.1 | 3 | 819 | |||
GenBank mRNA | BC040611.2 | - | 3 | 743 |

Gene Symbol: LINC01602 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130934.1 | 3 | 819 | |||
GenBank mRNA | BC040611.2 | - | 3 | 743 |