siRNA Details
Gene Symbol: LINC01556 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103538.1 | 1 | 717 | |||
GenBank mRNA | BX118328.1 | - | 1 | 717 |

Gene Symbol: LINC01556 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103538.1 | 1 | 717 | |||
GenBank mRNA | BX118328.1 | - | 1 | 717 |