siRNA Details
Gene Symbol: LINC00970 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104091.1 | 5 | 1098 | |||
GenBank mRNA | BC041008.1 | - | 5 | 1103 |

Gene Symbol: LINC00970 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104091.1 | 5 | 1098 | |||
GenBank mRNA | BC041008.1 | - | 5 | 1103 |