siRNA Details
Gene Symbol: LINC01405 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036513.1 | 1, 2 | 98 | |||
GenBank mRNA | AJ276555.1 | - | 1, 2 | 98 |

Gene Symbol: LINC01405 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036513.1 | 1, 2 | 98 | |||
GenBank mRNA | AJ276555.1 | - | 1, 2 | 98 |